Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cacodylic acid, sodium salt trihydrate, 98%, pure, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
500g
Packaging:
Glass bottle
6131-99-3
C2H12AsNaO5
214.024
MFCD00149079
RLGWPHBPRCROJO-UHFFFAOYSA-M
sodium cacodylate trihydrate,unii-r7a6nc7ygy,cacodylic acid sodium salt trihydrate,r7a6nc7ygy,dimethylarsonic acid sodium salt,cacodylic acid, sodium salt trihydrate,sodium dimethylarsinic acid trihydrate,dimethylarsenic acid sodium salt trihydrate,dimethylarsinic acid sodium salt trihydrate,hydroxydimethylarsine oxide sodium salt trihydrate
23679059
sodium;dimethylarsinate;trihydrate
C[As](=O)(C)[O-].O.O.O.[Na+]
6131-99-3
C2H12AsNaO5
214.024
MFCD00149079
RLGWPHBPRCROJO-UHFFFAOYSA-M
sodium cacodylate trihydrate,unii-r7a6nc7ygy,cacodylic acid sodium salt trihydrate,r7a6nc7ygy,dimethylarsonic acid sodium salt,cacodylic acid, sodium salt trihydrate,sodium dimethylarsinic acid trihydrate,dimethylarsenic acid sodium salt trihydrate,dimethylarsinic acid sodium salt trihydrate,hydroxydimethylarsine oxide sodium salt trihydrate
23679059
sodium;dimethylarsinate;trihydrate
C[As](=O)(C)[O-].O.O.O.[Na+]
Assay Percent Range 98%
Linear Formula (CH3)2As(O)ONa·3H2O
Merck Index 15, 8730
Formula Weight 214.03
Physical Form Crystals or Crystalline Powder
Percent Purity ≥97.5%
Grade Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 10g
Chemical Name or Material Cacodylic acid, sodium salt trihydrate, 98%
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
May cause cancer.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

RTECSNumber : CH7890000