Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACYBP Mouse anti-Human, Alexa Fluor™ 350, Clone: 3G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242677AF350

Catalog No. NP242677A30


Add to cart

Description

Description

CACYBP Monoclonal antibody specifically detects CACYBP in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

CACYBP
3G8
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
CacyBP, calcyclin binding protein, GIG5, hCacyBP, MGC87971, RP1-102G20.6, S100A6-binding protein, S100A6BPgrowth-inhibiting gene 5 protein, Siah-interacting protein, Siah-interacting protein (SIP), SIPcalcyclin-binding protein
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
CACYBP
Recombinant human CACYBP (1-185aa) purified from E. coli
100 μL
Primary
27101
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit