Learn More
Cadence Science™ Deflected Noncoring Septum Penetration Needles
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
Manufacturer:  Cadence Science™ 4174
 View more versions of this product
Catalog No. 14-825-15L
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
Stainless Steel
22 ga
51mm
Deflected non-coring septum
Deflected End
2 in.
Penetration Needle