Cadence Science™ Deflected Noncoring Septum Penetration Needles
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
Manufacturer: Cadence Science™ 4165
 View more versions of this product
Catalog No. 14-825-15B
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Gauge 26 ga
    Length (English) Needle 0.5 in.
    Length (Metric) Needle 12.7mm
    Type Penetration Needle
    For Use With (Application) Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
    Includes Deflected non-coring septum
    Material Stainless Steel
    Needle Point Style Deflected End
  • Description & Specifications
Related Products
Product Content Feedback