Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
For critical applications requiring sterile, flexible, single-use feeding needles
Manufacturer:  Cadence Science™ 9930
 View more versions of this product
Catalog No. 14-825-261
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
0.114 in.
Yes
4.7 in.
PTFE
For critical applications requiring sterile, flexible, single use feeding needles
0.029cm
14g
1.2cm
Yes
Soft, Flexible Animal feeding needle