Learn More
Cadence Science™ Hypodermic Needles
Manufacturer:  Cadence Science™ 4001/DELDISCBYVEND
 View more versions of this product
Catalog No. 14-825A
Description
Description
Slip-on, seamless tempered-steel needles with finished cutting edge, 12° regular medical point. In pasteboard boxes containing one dozen. For use with any BD™ disposable or reusable hypodermic syringe with Luer slip or needle lock tip.
Specifications
Specifications
Plated Brass (Hub)
0.01 in.
0.37 in.
12 Needles
Any BD disposable or reusable hypodermic syringe
Non-sterile
27 G
0.25 mm
Hypodermic Needle
12° Regular Medical
Luer