Learn More
Cadence Science™ Hypodermic Needles
Manufacturer:  Cadence Science™ 4019/DISCBYVEND
 View more versions of this product
Catalog No. 14-825C
Add to cart
Description
Description
Slip-on, seamless tempered-steel needles with finished cutting edge, 12° regular medical point. In pasteboard boxes containing one dozen. For use with any BD™ disposable or reusable hypodermic syringe with Luer slip or needle lock tip.
Specifications
Specifications
Plated Brass (Hub)
0.02 in.
0.62 in.
12 Needles
12° Regular Medical
Luer
25 G
0.5 mm
Hypodermic Needle
16 mm
Any BD disposable or reusable hypodermic syringe
Non-sterile