Learn More
Cadence Science™ Laboratory Pipetting Needles
Standard hub needles feature Type 304 stainless-steel cannulae for both manual and automatic volumetric dispensers, diluters, pipetters
Manufacturer:  Cadence Science™ 7936
 View more versions of this product
Catalog No. 14-825-16G
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
For both manual and automatic volumetric dispensers, diluters, pipetters and for laboratory procedures
101.6mm
Stainless steel
Luer
18 ga.
4 in.
Nickel Plated
90° blunt end
Pipetting Needle