Learn More
Cadence Science™ Micro-Emulsifying Needles
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
Manufacturer:  Cadence Science™ 7972
 View more versions of this product
Catalog No. 14-825-17D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar
2.87 in.
Glass
Micro Emulsifying Needle w/o Reinforcing Bar
Without reinforcing bar
7.3cm
Non-sterile
20 ga.