Learn More
Cadence Science™ Micro-Emulsifying Needles
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
Manufacturer:  Cadence Science™ 7974
 View more versions of this product
Catalog No. 14-825-17F
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar
Glass
25
7.3cm
Without reinforcing bar
No
2.87 in.
Micro Emulsifying Needle with Reinforcing Bar