Learn More
Cadence Science™ Non-Coring Septum Penetration Needles
Needles allow for easier penetration and minimize coring and tracking
Manufacturer:  Cadence Science™ 9862
 View more versions of this product
Catalog No. 01-290-28
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Needles allow for easier penetration and minimize coring and tracking
  • Micro-mate 316 stainless-steel hub
  • 316 stainless-steel tubing
Specifications
Specifications
20 G
1 in.
12 needles
25 mm
For easier penetration and minimize coring and tracking
Micro-Mate™ Stainless Steel
Penetration Needle
Deflected End