Cadence Science™ One-way Stopcocks
Precision-machined, individually lapped, and carefully mated to maintain pressure characteristics
Manufacturer: Cadence Science™ 6029
 View more versions of this product
Catalog No. 14-825-310
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Connectors Male luer-lock to 6mm hose end
    For Use With (Equipment) Other stopcocks, manifolds, adapters, tubing, needles and glass syringes
    Material Nickel Plated Brass
    Product Type 1-way Stopcock
  • Description & Specifications
Related Products
Product Content Feedback