Cadence Science™ One-way Stopcocks
Precision-machined, individually lapped, and carefully mated to maintain pressure characteristics
Manufacturer: Cadence Science™ 6027
 View more versions of this product
Catalog No. 14-825-313
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Connectors 8mm dia. hose end, 4mm dia. lumen
    For Use With (Equipment) Other stopcocks, manifolds, adapters, tubing, needles and glass syringes
    Material Nickel Plated Brass
    Product Type 1-way Stopcock
  • Description & Specifications
Related Products
Product Content Feedback