Learn More
Cadence Science™ One-way Stopcocks
Precision-machined, individually lapped, and carefully mated to maintain pressure characteristics
Manufacturer:  Cadence Science™ 6023
 View more versions of this product
Catalog No. 01-290-36
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Precision-machined, individually lapped, and carefully mated to maintain pressure characteristics
These stopcocks are easily integrated with other stopcocks, manifolds, adapters, tubing, needles, and glass syringes.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 PSI water pressure
  • High Pressure version is pressure resistant up to 20 bars (290 psi)
Specifications
Specifications
Nickel Plated Brass
1-way Stopcock
Male luer-lock to male luer-lock
Other stopcocks, manifolds, adapters, tubing, needles and glass syringes