Learn More
Cadence Science™ Perfektum™ Jumbo Glass Syringes
For biomedical, laboratory, clinical, research, and industrial applications
Manufacturer:  Cadence Science™ 5331
 View more versions of this product
Catalog No. 14-825-12E
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Jumbo Syringe
150mL
Individually
Each
Ideal for use in leak-resistant, high-flow rate medical and industrial applications, capable of dispensing large volumes of corrosive, toxic, and noxious liquids
150mL