Cadence Science™ Three-way Stopcocks
Manufacturer: Cadence Science™ 6010
 View more versions of this product
Catalog No. 14-630-101A
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Product Type 3-way Stopcock
    Connectors 1 female luer-lock, 1 male luer-lock and 1 right 3.5mm dia. hose end sidearm
    For Use With (Equipment) Other stopcocks, manifolds, adapters, tubing, needles and glass syringes
    Material Nickel Plated Brass
  • Description & Specifications
Related Products
Product Content Feedback