Learn More
Cadence Science™ Tuberculin Glass Syringes
Manufacturer:  Cadence Science™ 5200
 View more versions of this product
Catalog No. 14-826-100A
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Used in clinical and research procedures for the administration of precise lowvolume doses ranging from 0.25mL to 2mL
Borosilicate Glass
Tuberculin Syringe
0.01mL
Glass Luer Slip Tip
0.25mL