Learn More
Cadence Science™ Tuberculin Glass Syringes
Specifications
For Use With (Application) Used in clinical and research procedures for the administration of precise lowvolume doses ranging from 0.25mL to 2mL
Material Borosilicate Glass
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Graduations Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Graduations Price Quantity & Availability  
14-826-100A
View Documents
Cadence Science™
5200
0.25 mL 0.01 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
14-826-100B
View Documents
Cadence Science™
5201
0.5 mL 0.01 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Tuberculin mini glass syringes are widely used in clinical and research procedures for the administration of precise low volume doses ranging from 1/4mL to 2mL
  • Fabricated of chemically resistant borosilicate glass
  • Easy-to-read accurate permanent calibration lines
  • Glass Luer Slip Tip are reinforced glass bases to assure strong tips and minimize breakage
  • Tips have beveled ends to reduce chipping
Specifications
Specifications
Used in clinical and research procedures for the administration of precise lowvolume doses ranging from 0.25mL to 2mL
Borosilicate Glass