Learn More
Cadherin-13 Antibody (OTI3H6), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270385FR
Catalog No. NBP270385FR
Add to Cart
Description
Description
Cadherin-13 Monoclonal specifically detects Cadherin-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Cadherin-13
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
CDH13
Full length recombinant protein of human Cadherin-13 (NP_001248) produced in HEK293T cells.
0.1 mL
1012.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI3H6
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
cadherin 13, H-cadherin (heart), cadherin-13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, Truncated cadherin, truncated-cadherin
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Product Suggestions
Product Suggestions