Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-13 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems AF3264SP
This item is not returnable.
View return policy
Description
Cadherin-13 Polyclonal specifically detects Cadherin-13 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
Cadherin-13 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cadherin 13, H-cadherin (heart), cadherin-13, Cadherin13, CDH13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, T-Cadherin, Truncated cadherin, truncated-cadherin | |
Goat | |
Affinity Purified | |
RUO | |
1012 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
P55290 | |
CDH13 | |
Mouse myeloma cell line NS0-derived recombinant human Cadherin-13 Glu23-Ala692 Accession # P55290 | |
25 μg | |
Primary | |
Detects human Cadherin-13 in direct ELISAs and Western blots. In direct ELISAs, approximately 100% cross-reactivity with recombinant mouse Cadherin-13 is observed, and less than 10% cross-reactivity with recombinant human (rh) N-Cadherin is observed, and less than 2% cross-reactivity with rhCadherin-8, rhCadherin-11, rhCadherin-17, rhE-Cadherin, rhP-Cadherin, rhVE-Cadherin and rhR-Cadherin is observed. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only