Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-16 Antibody (CDH16/2125), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279856F
Description
Cadherin-16 Monoclonal specifically detects Cadherin-16 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cadherin-16 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CDH16 | |
Recombinant fragment (around aa 371-507) of human Cadherin-16 protein (exact sequence is proprietary) (Uniprot: O75309) | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CDH16/2125 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
Mouse | |
Protein A or G purified | |
RUO | |
This monoclonal antibody recognizes a protein of 130kDa, identified as Cadherin-16; Cadherin-16 has been suggested to distinguish Chromophobe Renal-Cell Carcinoma from Oncocytoma. | |
Store at 4°C in the dark. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction