Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-16 Mouse anti-Human, Clone: CDH16/2448, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30721310UG
Description
Cadherin-16 Monoclonal specifically detects Cadherin-16 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
Cadherin-16 | |
Monoclonal | |
0.2 mg/ml | |
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
Recombinant fragment (around aa 371-507) of human Cadherin-16 protein (exact sequence is proprietary) | |
100 μg | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
CDH16/2448 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
1014 | |
Store at 4°C. | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title