Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-16 Mouse anti-Human, Clone: CDH16/2448, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Cadherin-16
Clone CDH16/2448
Concentration 0.2 mg/ml
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications ELISA, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121899
View Documents
Novus Biologicals
NBP30721310UG
100 μg
Each for $352.89
Add to cart
 
NB121900
View Documents
Novus Biologicals
NBP30721320UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Cadherin-16
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
1014
IgG2b Kappa
Primary
Store at 4°C.
CDH16/2448
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Human
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin
Recombinant fragment (around aa 371-507) of human Cadherin-16 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit