Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-16 Mouse anti-Human, Mouse, Rat, Canine, Rabbit, Clone: rCDH16/1071, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen Cadherin-16
Clone rCDH16/1071
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121751
View Documents
Novus Biologicals
NBP30713910UG
100 μg
Each for $374.34
Add to cart
 
NB121752
View Documents
Novus Biologicals
NBP30713920UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Cadherin-16
0.2 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
1014
IgG1 Kappa
Primary
Store at 4°C.
rCDH16/1071
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Human, Mouse, Rat, Canine, Rabbit
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin
Recombinant full-length human Cadherin-16 protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit