Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-16 Rabbit anti-Human, Mouse, Rat, Allophycocyanin/Cy7, Clone: CDH16/1532R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254567APCCY7

Catalog No. N254567APC7


Add to cart

Description

Description

Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cadherin-16
CDH16/1532R
Flow Cytometry
Conjugated, Purified
CDH16
Recombinant human full-length CDH16 protein
Protein A purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse, Rat
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin
Rabbit
IgG Kappa
0.1 mL
Primary
1014.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit