Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-4/R-Cadherin Antibody (OTI4F5) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270384

Catalog No. NBP270384


Add to Cart

Description

Description

Cadherin-4/R-Cadherin Monoclonal specifically detects Cadherin-4/R-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Cadherin-4/R-Cadherin
Monoclonal
LYOPH
Western Blot 1:2000, Immunohistochemistry 1:150
CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin
Mouse
98.3 kDa
100 μg
1002
Human, Mouse, Rat
Lyophilized
Western Blot, Immunohistochemistry
OTI4F5
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
CDH4
Human recombinant protein fragment corresponding to amino acids 170-428 of human CDH4 (NP_001785) produced in E.coli.
Affinity Purified
Primary
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit