Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-4/R-Cadherin Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4F5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270384AF647

Catalog No. N270384A647


Add to cart

Description

Description

Cadherin-4/R-Cadherin Monoclonal antibody specifically detects Cadherin-4/R-Cadherin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Cadherin-4/R-Cadherin
OTI4F5
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CDH4
Recombinant protein fragment corresponding to amino acids 170-428 of human Cadherin-4/R-Cadherin (NP_001785) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin
Mouse
IgG1
0.1 mL
Primary
1002.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit