Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cadherin, Pan Ab-4, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cadherin, Pan Ab-4, Rabbit Polyclonal Antibody.

Specifications

Antigen Cadherin, Pan Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RB1524R1
View Documents
Thermo Scientific™
RB1524R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cadherin, Pan Ab-4, Rabbit Polyclonal Antibody.

Cadherins are calcium-dependent cell adhesion molecules, which play important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. They are expressed in a tissue-specific manner and are required for assembly of cells into solid tissue. Individual cadherin molecules are known to cooperate with each other to form a linear cell adhesion zipper. In adhesion junctions cadherins are bound to Beta and gamma catenins which in turn bind to alpha catenin, an actin binding protein. Cadherins play important part in tumor invasion and metastasis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Chicken, Dog, Monkey, Cow and Xenopus.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminal end of chicken N-cadherin

Positive Control: Tonsil, squamous epithelium

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cadherin, Pan Ab-4
Polyclonal
Rabbit
Junction Protein
A synthetic peptide from the C-terminal end of chicken N-cadherin
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit