Learn More
Abnova™ CADM1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00023705T02
Catalog No. 89-937-815
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CADM1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 42.7kDa
Transient overexpression cell lysate was tested with anti-CADM1 antibody (Mfr. No. H00023705-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
BC047021.1
pCMV-CADM1 full-length
100 μL
Human