Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CADM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118840
Description
Antibody detects endogenous levels of total CADM4.
CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).
Specifications
CADM4 | |
Polyclonal | |
Unconjugated | |
CADM4 | |
Cadm4; Cell adhesion molecule 4; Igdf4c; IGSF4C; Immunoglobulin superfamily member 4C; immunoglobulin superfamily, member 4C; membrane glycoprotein Tsll2; Necl4; NECL-4; nectin-like 4; nectin-like protein 4; SynCAM 4; synCAM4; TSLC1-like 2; TSLC1-like protein 2; Tsll2; Upar; urokinase plasminogen activator receptor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
199731, 260299, 365216 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q1WIM1, Q8NFZ8, Q8R464 | |
CADM4 | |
A synthesized peptide derived from human CADM4(Accession Q8NFZ8), corresponding to amino acid residues L356-I388. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction