Alfa Aesar™ Cadmium 2,4-pentanedionate, 98%

Manufacturer: Alfa Aesar™ 4196806

Quantity 5g

 View more versions of this product

Catalog No. AA4196806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14689-45-3
    Formula Weight 310.64
    MDL Number MFCD00040412
    UN Number UN2570
    Synonym Cadmium acetylacetonate
    Chemical Name or Material Cadmium 2,4-pentanedionate

    Specifications

    Formula Weight 310.64
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Cadmium acetylacetonate
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
    Quantity 5g
    Physical Form Powder
    MDL Number MFCD00040412
    UN Number UN2570
    CAS 14689-45-3
    Health Hazard 2 GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    RTECS Number EV0260000
    Chemical Name or Material Cadmium 2,4-pentanedionate
    Assay 98%

    GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit