Alfa Aesar™ Cadmium acetate dihydrate, 98%

In cadium electroplating

Manufacturer: Alfa Aesar™ 4158122

Quantity 100g

 View more versions of this product

Catalog No. AA4158122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5743-04-4
    Formula Weight 266.54 (230.50 Anhydrous)
    MDL Number MFCD00012449
    UN Number UN2570
    Merck Index 14,1614
    Chemical Name or Material Cadmium acetate dihydrate

    Specifications

    Formula Weight 266.54 (230.50 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00012449
    UN Number UN2570
    Quantity 100g
    Solubility Information Freely soluble in water, soluble in alcohol
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
    RTECS Number AF7505000
    TSCA TSCA
    Physical Form Powder
    Sensitivity Hygroscopic
    Merck Index 14,1614
    Color White
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Density 2.01g/mL
    CAS 5743-04-4
    Chemical Name or Material Cadmium acetate dihydrate
    Assay 98%

    GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit