Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium Acetate, Dihydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™
Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
Specifications
Formula Weight | 266.52 |
Physical Form | Crystals |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.01kg |
Quantity | 500g |
Chemical Name or Material | Cadmium Acetate, Dihydrate |