Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Acetate, Dihydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
5743-04-4
C4H10CdO6
266.53
AUIZLSZEDUYGDE-UHFFFAOYSA-L
5743-04-4
C4H10CdO6
266.53
AUIZLSZEDUYGDE-UHFFFAOYSA-L

High quality chemicals for laboratory and specialized industrial use

Formula Weight 266.52
Physical Form Crystals
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.01kg
Quantity 125g
Chemical Name or Material Cadmium Acetate, Dihydrate