Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Cadmium Acetate Dihydrate, purum p.a., ≥98.0% (KT), Honeywell™

Click to view available options
Quantity:
10g
100g
500g
Packaging:
Poly Bottle
5743-04-4
C4H10CdO6
266.53
MFCD00150020
AUIZLSZEDUYGDE-UHFFFAOYSA-L
bis 1-hydroxycyclohexyl butadiyne,1,4-bis 1-hydroxycyclohexyl-1,3-butadiyne,cyclohexanol, 1,1'-butadiynylenedi,1,1'-butadiynylenedicyclohexanol,cyclohexanol, 1,1'-1,3-butadiyne-1,4-diyl bis,1-4-1-hydroxycyclohexyl buta-1,3-diyn-1-yl cyclohexan-1-ol,acmc-1aky0,cambridge id 5106496,cyclohexanol,1'-butadiynylenedi,6-aminobenzoic acid propyl ester
79835
1-[4-(1-hydroxycyclohexyl)buta-1,3-diynyl]cyclohexan-1-ol
O.O.[Cd++].CC([O-])=O.CC([O-])=O
5743-04-4
C4H10CdO6
266.53
MFCD00150020
AUIZLSZEDUYGDE-UHFFFAOYSA-L
bis 1-hydroxycyclohexyl butadiyne,1,4-bis 1-hydroxycyclohexyl-1,3-butadiyne,cyclohexanol, 1,1'-butadiynylenedi,1,1'-butadiynylenedicyclohexanol,cyclohexanol, 1,1'-1,3-butadiyne-1,4-diyl bis,1-4-1-hydroxycyclohexyl buta-1,3-diyn-1-yl cyclohexan-1-ol,acmc-1aky0,cambridge id 5106496,cyclohexanol,1'-butadiynylenedi,6-aminobenzoic acid propyl ester
79835
1-[4-(1-hydroxycyclohexyl)buta-1,3-diynyl]cyclohexan-1-ol
O.O.[Cd++].CC([O-])=O.CC([O-])=O

Purum p.a.,≥98.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Linear Formula Cd(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3730567
Formula Weight 266.53g/mol
Percent Purity ≥98.0% (KT)
Grade purum p.a.
Density 2.01g/cu.cm
Packaging Poly Bottle
Physical State Solid
Melting Point 256°C
Quantity 100g
Chemical Name or Material Cadmium Acetate Dihydrate
Show More Show Less

P201-P261-P273-P280-P308 + P313-P501

H302 + H312 + H332-H350-H410

DOTInformation : 9/PGIII

EINECSNumber : 208-853-2

RTECSNumber : AF7505000