Alfa Aesar™ Cadmium arsenide, 99% (metals basis)

Manufacturer: Alfa Aesar™ 2277318

Quantity 50g

 View more versions of this product

Catalog No. AA2277318

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12006-15-4
    Linear Formula Cd3As2
    Formula Weight 487.04
    MDL Number MFCD00049723
    UN Number UN1557
    Chemical Name or Material Cadmium arsenide

    Specifications

    Physical Form Lumps
    Formula Weight 487.04
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    MDL Number MFCD00049723
    Color Dark gray
    Quantity 50g
    Health Hazard 1 Danger
    Linear Formula Cd3As2
    Density 3.031g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
    TSCA TSCA
    CAS 12006-15-4
    UN Number UN1557
    Health Hazard 2 GHS H Statement
    H300-H330-H350
    Fatal if swallowed.
    Fatal if inhaled.
    May cause cancer.
    Chemical Name or Material Cadmium arsenide
    Assay 99%, (metals basis)

    GHS H Statement
    H300-H330-H350
    Fatal if swallowed.
    Fatal if inhaled.
    May cause cancer.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit