Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cadmium bromide hydrate, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4449814

Quantity 25g

 View more versions of this product

Catalog No. AA4449814

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7789-42-6
272.22 (Anhydrous)
UN2570
CdBr2·xH2O
MFCD00150204
Cadmium bromide hydrate

Specifications

7789-42-6
CdBr2·xH2O
MFCD00150204
Odorless
White to pale yellow
Cadmium bromide hydrate
Crystalline
272.22 (Anhydrous)
UN2570
Hygroscopic
25g
99.999%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

RTECSNumber : EU9935000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit