Alfa Aesar™ Cadmium bromide, ultra dry, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 3568203

Quantity 1g

 View more versions of this product

Catalog No. AA3568203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-42-6
    Formula Weight 272.22
    MDL Number MFCD00015996
    UN Number UN2570
    Merck Index 14,1615
    Chemical Name or Material Cadmium bromide, Ultra dry

    Specifications

    Formula Weight 272.22
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Odor Odorless
    CAS 7789-42-6
    UN Number UN2570
    Merck Index 14,1615
    Color White to pale yellow
    Quantity 1g
    RTECS Number EU9935000
    TSCA TSCA
    Mesh Size −10 Mesh
    Physical Form Beads
    Sensitivity Hygroscopic
    MDL Number MFCD00015996
    Boiling Point 863°C
    Melting Point 567°C
    Health Hazard 1 Warning
    Storage Note 1
    • Ampouled under argon
    Density 5.192g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
    Chemical Name or Material Cadmium bromide, Ultra dry
    Assay 99.999%, (metals basis)

    GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit