Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium bromide, ultra dry, 99.999% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Specifications
Boiling Point | 863°C |
Melting Point | 567°C |
Color | White to Yellow |
Physical Form | Powder |
Quantity | 5 g |
UN Number | UN2570 |
Merck Index | 14,1615 |
Solubility Information | Soluble in alcohol,ether,acetone and liquid ammonia. |
Formula Weight | 272.22 |
Percent Purity | 99.999% |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H350 |
Hazard Statement | GHS H Statement H302-H312-H332 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: CADMIUM COMPOUNDS |
EINECSNumber | 232-165-1 |
RTECSNumber | EU9935000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures; Store under Nitrogen |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction