Cadmium carbonate, >99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 12229A1

Quantity 1kg

 View more versions of this product

Catalog No. AA12229A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 513-78-0
    Physical Form Powder
    Assay Percent Range >99% (Metal basis)
    Molecular Formula CdCO3
    Formula Weight 172.42
    MDL Number MFCD00010918
    Chemical Name or Material Cadmium carbonate

    Specifications

    Formula Weight 172.42
    Quantity 1kg
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 208-168-9
    Physical Form Powder
    Assay Percent Range >99% (Metal basis)
    CAS 513-78-0
    Odor Odorless
    Molecular Formula CdCO3
    Appearance White powder
    MDL Number MFCD00010918
    Health Hazard 1 H400-H410-H302-H312-H332
    Chemical Name or Material Cadmium carbonate

    H400-H410-H302-H312-H332

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit