Learn More
Cadmium Carbonate, Powder, Reagent, 97%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Bottle
C1020, 513-78-0, CdCO3
Spectrum™ Cadmium Carbonate, Powder, Reagent , also known as Otavite, a naturally occurring mineral. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Lead (Pb) 1e-005
Loss on Ignition 25.5-27.5%
Packaging Poly Bottle