Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Carbonate, Powder, Reagent, 97%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle
513-78-0
CCdO3
172.42
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium(2+) carbonate
[Cd++].[O-]C([O-])=O
513-78-0
CCdO3
172.42
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium(2+) carbonate
[Cd++].[O-]C([O-])=O

C1020, 513-78-0, CdCO3

Spectrum™ Cadmium Carbonate, Powder, Reagent , also known as Otavite, a naturally occurring mineral. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
CAS Max % 100%
UN Number UN2570
Percent Purity 97%
Lead (Pb) 0.001%
Loss on Ignition 25.5-27.5%
Packaging Fiber Drum
Zinc (Zn) 0.05%
Quantity 50 kg