Cadmium carbonate, White powder, 99.998% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1066018

Quantity 50g

 View more versions of this product

Catalog No. AA1066018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 513-78-0
    Physical Form Powder
    Assay Percent Range 99.998% (Metals basis)
    Molecular Formula CdCO3
    Formula Weight 172.42
    MDL Number MFCD00010918
    Chemical Name or Material Cadmium carbonate

    Specifications

    Physical Form Powder
    Formula Weight 172.42
    Quantity 50g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    Odor Odorless
    Assay Percent Range 99.998% (Metals basis)
    Molecular Formula CdCO3
    MDL Number MFCD00010918
    CAS 513-78-0
    Health Hazard 1 H400-H410-H302-H312-H332: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
    Health Hazard 2 This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets
    Chemical Name or Material Cadmium carbonate

    This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets

    H400-H410-H302-H312-H332: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit