Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium carbonate, White powder, 99.998% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1066009

Quantity 10g

 View more versions of this product

Catalog No. AA1066009


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

513-78-0
99.998% (Metals basis)
172.42
Cadmium carbonate
Powder
CdCO3
MFCD00010918

Specifications

513-78-0
99.998% (Metals basis)
172.42
4.26g/cm3
White
Cadmium carbonate
Powder
CdCO3
MFCD00010918
Odorless
10g

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312-H332

GHS P Statement
P261-P280-P264-P270-P304+P340-P301+P312-P312-P363-P322-P501a

Warning

DOTInformation : DOT Class: 6.1, Packaging Group: III

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit