Cadmium chloride hemipentahydrate, ACS, 79.5 to 81.0%, Alfa Aesar™

Manufacturer: Alfa Aesar 1237318

Quantity 50g

 View more versions of this product

Catalog No. AA1237318

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 233-296-7
    Physical Form Crystalline
    Assay Percent Range 79.5-81.0%
    Molecular Formula CdCl2·2.5H2O
    Formula Weight 228.36
    MDL Number MFCD00799071
    Chemical Name or Material Cadmium chloride hemipentahydrate
    Grade ACS Grade

    Specifications

    Formula Weight 228.36
    Quantity 50g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 233-296-7
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range 79.5-81.0%
    Molecular Formula CdCl2·2.5H2O
    Appearance White crystalline
    MDL Number MFCD00799071
    CAS 233-296-7
    Health Hazard 1 H301-H330-H340-H350-H360-H372-H400-H410
    Chemical Name or Material Cadmium chloride hemipentahydrate
    Grade ACS Grade

    H301-H330-H340-H350-H360-H372-H400-H410

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit