Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium chloride hemipentahydrate, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 34.0°C |
Color | White |
Physical Form | Crystals and/or Chunks |
Quantity | 100 g |
Linear Formula | CdCl2·5/2H2O |
Merck Index | 15, 1622 |
Solubility Information | Solubility in water: 1680g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in ethanol and methanol |
Formula Weight | 228.34 |
Percent Purity | 79.5 to 81.0% (CdCl2) |
Grade | ACS Reagent |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause cancer. May cause genetic defects. May damage fertility. May damage the unborn child. Fatal if inhaled. Toxic if swallowed. Causes damage to organs through prolonged or repeated exposure |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. Avoid release to the environment. IF INHALED: Remove victim to fresh air and keep at rest in a positi |
RTECSNumber | EV0178000 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction