Alfa Aesar™ Cadmium chloride, ultra dry, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 3572809

Quantity 10g

 View more versions of this product

Catalog No. AA3572809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10108-64-2
    Formula Weight 183.32
    MDL Number MFCD00010916
    UN Number UN2570
    Merck Index 14,1617
    Chemical Name or Material Cadmium chloride, Ultra dry

    Specifications

    Formula Weight 183.32
    Packaging Ampule
    Health Hazard 2 GHS H Statement
    H301-H330-H340-H350-H360-H372
    Toxic if swallowed.
    Fatal if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −10 Mesh
    Odor Odorless
    Sensitivity Hygroscopic
    CAS 10108-64-2
    Merck Index 14,1617
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
    Physical Form Beads
    UN Number UN2570
    Boiling Point 960°C
    Melting Point 568°C
    Health Hazard 1 Danger
    Density 4.047g/mL
    TSCA TSCA
    MDL Number MFCD00010916
    Quantity 10g
    Storage Note 1
    • Ampouled under argon
    RTECS Number EV0175000
    Chemical Name or Material Cadmium chloride, Ultra dry
    Assay 99.9%, (metals basis)

    GHS H Statement
    H301-H330-H340-H350-H360-H372
    Toxic if swallowed.
    Fatal if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit