Alfa Aesar™ Cadmium cyclohexanebutyrate

As an organophile Cadmium compound

Manufacturer: Alfa Aesar™ 4197003

Quantity 1g

 View more versions of this product

Catalog No. AA4197003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 55700-14-6
    Formula Weight 450.9
    MDL Number MFCD00036398
    UN Number UN2570
    Synonym Cyclohexanebutyric acid cadmium salt; 4-Cyclohexylbutyric acid cadmium salt
    Chemical Name or Material Cadmium cyclohexanebutyrate

    Specifications

    Formula Weight 450.9
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Physical Form Powder
    UN Number UN2570
    Synonym Cyclohexanebutyric acid cadmium salt; 4-Cyclohexylbutyric acid cadmium salt
    Health Hazard 1 Warning
    Quantity 1g
    TSCA TSCA
    MDL Number MFCD00036398
    CAS 55700-14-6
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
    Chemical Name or Material Cadmium cyclohexanebutyrate

    GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit