Alfa Aesar™ Cadmium fluoride, Puratronic™, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4020314

Quantity 25g

 View more versions of this product

Catalog No. AA4020314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-79-6
    Formula Weight 150.4
    MDL Number MFCD00010919
    UN Number UN2570
    Merck Index 14,1619
    Chemical Name or Material Cadmium fluoride
    Grade Puratronic™

    Specifications

    Formula Weight 150.4
    Health Hazard 2 GHS H Statement
    H301-H330-H340-H350-H360-H372
    Toxic if swallowed.
    Fatal if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2570
    Odor Odorless
    Merck Index 14,1619
    Melting Point 1100°C
    Health Hazard 1 Danger
    Physical Form Powder
    CAS 7790-79-6
    Boiling Point 1758°C
    Quantity 25g
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CADMIUM COMPOUNDS
    TSCA TSCA
    MDL Number MFCD00010919
    Density 6.64g/mL
    Chemical Name or Material Cadmium fluoride
    Assay 99.99%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H301-H330-H340-H350-H360-H372
    Toxic if swallowed.
    Fatal if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit