Alfa Aesar™ Cadmium hydroxide, Cd 75% min

Cadmium plating, storage battery electrodes

Manufacturer: Alfa Aesar™ 8929722

Quantity 100g

 View more versions of this product

Catalog No. AA8929722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21041-95-2
    Molecular Formula Cd(OH)2
    Formula Weight 146.41
    MDL Number MFCD00015995
    UN Number UN2570
    Merck Index 14,1620
    Chemical Name or Material Cadmium hydroxide

    Specifications

    Formula Weight 146.41
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Density 4.79g/mL at 15°C
    RTECS Number EV1260000
    Physical Form Crystalline
    Quantity 100g
    Concentration Cd 75% min
    Sensitivity Air Sensitive
    Molecular Formula Cd(OH)2
    MDL Number MFCD00015995
    CAS 21041-95-2
    UN Number UN2570
    Merck Index 14,1620
    Melting Point 300°C (dec.)
    Solubility Information Soluble in dilute acids, NH4Cl solutions
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
    Chemical Name or Material Cadmium hydroxide

    GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit